Learn More
Electron Microscopy Sciences Easy-Load Cover for Slotted Cassettes, Gray
SDP
Catalog No. 5030302
Encompass_Preferred
Easy-Load Slotted Cassettes one-piece disposable plastic lid can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: Gray
Catalog No. 50-303-02 Supplier Electron Microscopy Sciences Supplier No. 70061GR
Easy-Load Slotted Cassettes one-piece disposable plastic lid can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: Gray