Learn More
Electron Microscopy Sciences Easy-Load Cover for Slotted Cassettes, Peach
SDP
Catalog No. 5030311
Encompass_Preferred
Easy-Load Slotted Cassettes one-piece disposable plastic lid can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: Peach
Catalog No. 50-303-11 Supplier Electron Microscopy Sciences Supplier No. 70061PE
Easy-Load Slotted Cassettes one-piece disposable plastic lid can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: Peach