Learn More
Electron Microscopy Sciences Easy-Load Cover for Slotted Cassettes, Yellow
SDP
Catalog No. 5030310
Encompass_Preferred
One-piece disposable plastic, it can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: Yellow
Catalog No. 50-303-10 Supplier Electron Microscopy Sciences Supplier No. 70061Y
One-piece disposable plastic, it can be opened and closed as often as necessary and it always relocks securely without danger of specimen loss.
Color: Yellow