Learn More
IMEB, Inc Tek-Select Cover Glass, #1 Thickness, 24x50, 10/Case, Quality Cover Glass Offer Superior Resistance to Corrosion, Thickness and Flatness Are Uniform, Free From Striae, Oily Films and Fingerprints
SDP
Catalog No. 501689500
Encompass_Preferred
Tek-Select Cover Glass, #1 Thickness, 24x50, 10 per Case. Our quality cover glass offer superior resistance to corrosion. Thickness and flatness are uniform. Cover glass are completely free from striae, oily films and fingerprints. They are packed in a clear, rigid styrene box with semisoft, lint-free foam interior. Foil lining keeps product, dust and moisture FREE. #1 thickness; 0.13-0.17mm.
Catalog No. 50-168-9500 Supplier IMEB, Inc Supplier No. CG12450
Tek-Select Cover Glass, #1 Thickness, 24x50, 10 per Case. Our quality cover glass offer superior resistance to corrosion. Thickness and flatness are uniform. Cover glass are completely free from striae, oily films and fingerprints. They are packed in a clear, rigid styrene box with semisoft, lint-free foam interior. Foil lining keeps product, dust and moisture FREE. #1 thickness; 0.13-0.17mm.