Learn More
THORLABS INC Precision Cover Glasses, #1.5H Thickness, Ø12 mm | Pack of 1000
Catalog No. NC1692862
Encompass
Thorlabs' Precision Cover Glasses are ground to a uniform thickness of 170 ± 5 µm and are made from Schott D 263 M Glass, an optically clear and colorless glass that exhibits low auto-fluorescence. The uniform thickness and consistent optical properties help reduce image defects due to spherical aberrations (see the Specs tab for details). As such, these cover slips are ideal for use with high NA objective lenses.
Catalog No. NC1692862 Supplier THORLABS INC Supplier No. CG15NH
Thorlabs' Precision Cover Glasses are ground to a uniform thickness of 170 ± 5 µm and are made from Schott D 263 M Glass, an optically clear and colorless glass that exhibits low auto-fluorescence. The uniform thickness and consistent optical properties help reduce image defects due to spherical aberrations (see the Specs tab for details). As such, these cover slips are ideal for use with high NA objective lenses.