Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1.5, 22x22 mm, 138 per oz 1 OZ
SDP
Catalog No. 50365603
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 138 per oz
Thickness: .16 - .19m
Catalog No. 50-365-603 Supplier Electron Microscopy Sciences Supplier No. 7220401
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 138 per oz
Thickness: .16 - .19m