Learn More
Carolina Science & Math Carolina Cover Glasses, Circle, 12 mm, Thickness, 0.09 to 0.12mm, Box of 100
SDP
Catalog No. NC9216450
Encompass_Preferred
Cover Glass; Circles; Carolina;; High quality borosilicate glass of the highest hydrolytic consistency for fluorescent microscopy; 12mm; 0.09 to 0.12mm T; Deutsche Spiegelglas micro sheet; 100/BX
Catalog No. NC9216450 Supplier Carolina Science & Math Supplier No. 633009
Cover Glass; Circles; Carolina;; High quality borosilicate glass of the highest hydrolytic consistency for fluorescent microscopy; 12mm; 0.09 to 0.12mm T; Deutsche Spiegelglas micro sheet; 100/BX