Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #0, 22x22 mm, 208 per oz 1 OZ
SDP
Catalog No. 501899777
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 208 per oz
Thickness: .08 - .13mm
Catalog No. 50-189-9777 Supplier Electron Microscopy Sciences Supplier No. 7219810
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 208 per oz
Thickness: .08 - .13mm