Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1, 22x22 mm, 156 per oz 1 OZ
SDP
Catalog No. 501899778
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 156 per oz
Thickness: .13 - .17mm
Catalog No. 50-189-9778 Supplier Electron Microscopy Sciences Supplier No. 7220010
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 156 per oz
Thickness: .13 - .17mm