Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #2, 22x22 mm, 116 per oz 1 OZ
SDP
Catalog No. 501899791
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 116 per oz
Thickness: .17 - .25mm
Catalog No. 50-189-9791 Supplier Electron Microscopy Sciences Supplier No. 7221010
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 22 mm
Package: 116 per oz
Thickness: .17 - .25mm