Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1, 22x30 mm, 116 per oz 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7220020
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 30 mm
Package: 116 per oz
Thickness: .13 - .17mm
Catalog No. 50-189-9780