Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1, 22x40 mm, 87 per oz 1 OZ
SDP
Catalog No. 501899782
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 40 mm
Package: 87 per oz
Thickness: .13 - .17mm
Catalog No. 50-189-9782 Supplier Electron Microscopy Sciences Supplier No. 7220030
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 40 mm
Package: 87 per oz
Thickness: .13 - .17mm