Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1, 22x40 mm, 87 per oz 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7220031
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 40 mm
Package: 87 per oz
Thickness: .13 - .17mm
Catalog No. 50-189-9783