Learn More
Electron Microscopy Sciences Microscope Slide Cover Glass #1.5, 22x50 mm, 102 per oz 1 OZ
SDP
Catalog No. 501899786
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 50 mm
Package: 102 per oz
Thickness: .16 - .19m
Catalog No. 50-189-9786 Supplier Electron Microscopy Sciences Supplier No. 7220404
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Size: 22 x 50 mm
Package: 102 per oz
Thickness: .16 - .19m