Learn More
Ted Pella Inc Cover Glass, 50 x 75mm (1.96 x 2.95"), Thickness No. 1, 100/pk
SDP
Catalog No. NC1784030
Encompass
Made of optical grade borosilicate glass. Made for the large microscope slides of 3" x 2" or larger.
Catalog No. NC1784030 Supplier Ted Pella Inc Supplier No. 260462100
Made of optical grade borosilicate glass. Made for the large microscope slides of 3" x 2" or larger.