Learn More
Ted Pella Inc Coverglass for Super Mega Slides, #1.5 Thickness, 64 x 50mm, 100/pk
SDP
Catalog No. NC1708519
Encompass
This coverglass is sized to match the working area of the HISTOBOND + Super Mega Slide. Thickness: #1.5, Dimensions: 64 x 50mm
Catalog No. NC1708519 Supplier Ted Pella Inc Supplier No. 260378
This coverglass is sized to match the working area of the HISTOBOND + Super Mega Slide. Thickness: #1.5, Dimensions: 64 x 50mm