Learn More
BRAIN RESEARCH LABORATORIES Brain Research Laboratories Cover Glass, 50 mm x 75 mm, No. 1 Thickness, 100 pcs
Catalog No. NC1752532
Encompass
Discover the Brain Research Laboratories Cover Glass, a premium 50 mm x 75 mm product crafted from high-quality Schott D 263 M borosilicate glass. Engineered for microscopy, it provides excellent optical clarity and chemical resistance, ensuring reliable research results. Ideal for light and fluorescence microscopy, its precise thickness minimizes optical aberrations, setting it apart from ordinary glass.
Catalog No. NC1752532 Supplier BRAIN RESEARCH LABORATORIES Supplier No. 50751D
Discover the Brain Research Laboratories Cover Glass, a premium 50 mm x 75 mm product crafted from high-quality Schott D 263 M borosilicate glass. Engineered for microscopy, it provides excellent optical clarity and chemical resistance, ensuring reliable research results. Ideal for light and fluorescence microscopy, its precise thickness minimizes optical aberrations, setting it apart from ordinary glass.