Learn More
Electron Microscopy Sciences Circular Cover Glass #1.5, 12 mm, 692 Pieces/oz 1 OZ
SDP
Catalog No. 50949054
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 12 mm
Package: 692 Pieces/oz
Thickness: 0.16-0.19 mm
Catalog No. 50-949-054 Supplier Electron Microscopy Sciences Supplier No. 7223110
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 12 mm
Package: 692 Pieces/oz
Thickness: 0.16-0.19 mm