Learn More
Electron Microscopy Sciences Circular Cover Glass #1, 15 mm, 443 Pieces/oz 1 OZ
SDP
Catalog No. 50949051
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 15 mm
Package: 443 Pieces/oz
Thickness: 0.13-0.17 mm
Catalog No. 50-949-051 Supplier Electron Microscopy Sciences Supplier No. 7222810
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 15 mm
Package: 443 Pieces/oz
Thickness: 0.13-0.17 mm