Learn More
Electron Microscopy Sciences Circular Cover Glass #1, 18 mm, 308 Pieces/oz 1 OZ
SDP
Catalog No. 50949048
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 18 mm
Package: 308 Pieces/oz
Thickness: 0.13-0.17 mm
Catalog No. 50-949-048 Supplier Electron Microscopy Sciences Supplier No. 7222910
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 18 mm
Package: 308 Pieces/oz
Thickness: 0.13-0.17 mm