Learn More
Electron Microscopy Sciences Circular Cover Glass #1, 22 mm, 206 Pieces/oz 1 OZ
SDP
Catalog No. 50949044
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 22 mm
Package: 206 Pieces/oz
Thickness: 0.13-0.17 mm
Catalog No. 50-949-044 Supplier Electron Microscopy Sciences Supplier No. 7222110
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 22 mm
Package: 206 Pieces/oz
Thickness: 0.13-0.17 mm