Learn More
Electron Microscopy Sciences Circular Cover Glass #1, 25 mm, 159 Pieces/oz 1 OZ
SDP
Catalog No. 50948978
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 25 mm
Package: 159 Pieces/oz
Thickness: 0.13-0.17 mm
Catalog No. 50-948-978 Supplier Electron Microscopy Sciences Supplier No. 7222301
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 25 mm
Package: 159 Pieces/oz
Thickness: 0.13-0.17 mm