Learn More
Electron Microscopy Sciences Circular Cover Glass #1.5, 18 mm, 266 Pieces/oz 1 OZ
SDP
Catalog No. 50948975
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 18 mm
Package: 266 Pieces/oz
Thickness: 0.16-0.19 mm
Catalog No. 50-948-975 Supplier Electron Microscopy Sciences Supplier No. 7220041
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 18 mm
Package: 266 Pieces/oz
Thickness: 0.16-0.19 mm