Learn More
Electron Microscopy Sciences Circular Cover Glass #1.5, 22 mm, 178 Pieces/oz 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7222410
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 22 mm
Package: 178 Pieces/oz
Thickness: 0.16-0.19 mm
Catalog No. 50-949-047