Learn More
Electron Microscopy Sciences Circular Cover Glass #2, 12 mm, 502 Pieces/oz 1 OZ
SDP
Supplier:  Electron Microscopy Sciences 7222610
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 12 mm
Package: 502 Pieces/oz
Thickness: 0.17-0.25 mm
Catalog No. 50-949-052