Learn More
Electron Microscopy Sciences Circular Cover Glass #2, 12 mm, 502 Pieces/oz 1 OZ
SDP
Catalog No. 50949005
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 12 mm
Package: 502 Pieces/oz
Thickness: 0.17-0.25 mm
Catalog No. 50-949-005 Supplier Electron Microscopy Sciences Supplier No. 7222601
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 12 mm
Package: 502 Pieces/oz
Thickness: 0.17-0.25 mm