Learn More
Electron Microscopy Sciences Circular Cover Glass #2, 15 mm, 321 Pieces/oz 1 OZ
SDP
Catalog No. 50949045
Encompass_Preferred
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 15 mm
Package: 321 Pieces/oz
Thickness: 0.17-0.25 mm
Catalog No. 50-949-045 Supplier Electron Microscopy Sciences Supplier No. 7222710
Manufactured from clear white, borosilicate glass. They offer super clarity, are perfectly flat, and corrosion resistant. All cover glasses are uniform in size and thickness.
Diameter: 15 mm
Package: 321 Pieces/oz
Thickness: 0.17-0.25 mm