Learn More
Electron Microscopy Sciences COVER GLASS NO1.5 22X50MM
SDP
Catalog No. NC2458520
Encompass_Preferred
Made of chemically resistant borosilicate glass D 263®M of the first hydrolytic classAbsolutely colorless, perfectly clear, suitable for fluorescence microscopyIn compliance with DIN ISO 8255For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark and batch number for comprehensive information and traceability.
Catalog No. NC2458520 Supplier Electron Microscopy Sciences Supplier No. 71861068C
Made of chemically resistant borosilicate glass D 263®M of the first hydrolytic classAbsolutely colorless, perfectly clear, suitable for fluorescence microscopyIn compliance with DIN ISO 8255For in-vitro diagnostic (IVD) applications according to IVD directive 98/79/EC, with CE-mark and batch number for comprehensive information and traceability.