Learn More
Andwin Scientific COVER GLASS NO1.5 24X60MM
SDP
Catalog No. NC9008898
Encompass_Preferred
The Gold Seal cover glass is manufactured from pre-selected, pre-cleaned silicate glass. This product has a standard size of 24 x 60 mm and has a thickness of 0.16-0.19 mm.
Catalog No. NC9008898 Supplier Andwin Scientific Supplier No. 6379301
The Gold Seal cover glass is manufactured from pre-selected, pre-cleaned silicate glass. This product has a standard size of 24 x 60 mm and has a thickness of 0.16-0.19 mm.