Learn More
Electron Microscopy Sciences Large Coverglass Staining Rack, Stainless Steel
SDP
Catalog No. 50949590
Encompass_Preferred
Made from polished stainless steel, this staining rack securely holds thirty 22 or 24 mm cover glass. Welded stainless steel. Spring-type detachable handle. 4" (10.2cm) long.
Catalog No. 50-949-590 Supplier Electron Microscopy Sciences Supplier No. 7223904
Made from polished stainless steel, this staining rack securely holds thirty 22 or 24 mm cover glass. Welded stainless steel. Spring-type detachable handle. 4" (10.2cm) long.