Learn More
Cole-Parmer Cole-Parmer Cover glasses, 22 x 22 mm square, 0.13 to 0.17 mm thickness
Catalog No. NC1173827 Shop All Cole-Parmer Products
Encompass_Preferred
Cover glasses are made from borosilicate glass. Square cover glass is 22 x 22 mm with a thickness of #1 (0.13 to 0.17 mm).
Catalog No. NC1173827 Supplier Cole-Parmer Supplier No. 4851000
Cover glasses are made from borosilicate glass. Square cover glass is 22 x 22 mm with a thickness of #1 (0.13 to 0.17 mm).