Learn More
Hunter Lab COVER OPAQUE FOR SAMPLE CUP
SDP
Catalog No. NC0290890
Encompass
Provides a light trap to exclude the interference of external ambient light on the sample
Catalog No. NC0290890 Supplier Hunter Lab Supplier No. 04400000
Provides a light trap to exclude the interference of external ambient light on the sample