Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Cover Plates
SCI_ED

Catalog No. S24591 Shop All GSC Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Dimensions (L x W):
3 x 3 in.
4.5 x 4.5 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S24591 3 x 3 in.
S24590 4.5 x 4.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24591 Supplier GSC Go Science Crazy Supplier No. GP312
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to cover beaker and flasks. GSC International Cover Plates are made of glass.

  • Features ground edges
  • Autoclavable
  • Keeps the content of flask free from contaminants
  • Available in different sizes

Specifications

Description Cover Plate
Material Glass
Shape Square
Type Cover Plate
Dimensions (L x W) 3 x 3 in.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.