Learn More
Med Vet International Ultrasound Probe Cover Eclipse 1-3/4 X 9-1/2 inch Polyisoprene Nonsterile For Use With Ultrasound Endocavity Probe, 38-01, 100/Box
Catalog No. 502829749
Encompass_Preferred
Pre-gelled inside with Aquasonic 100 ultrasound transmission gel Only one probe cover is required for each procedure Fits most endocavity ultrasound probes
Catalog No. 50-282-9749 Supplier Med Vet International Supplier No. 316682BX
Pre-gelled inside with Aquasonic 100 ultrasound transmission gel Only one probe cover is required for each procedure Fits most endocavity ultrasound probes