Learn More
Med Vet International Tympanic Thermometer Probe Cover Thermoscan For Use With Thermoscan Thermometer, 06000-005, 1/Box
Catalog No. 502832159
Encompass_Preferred
Thermoscan Pro 6000 probe covers are compatible for use with Thermoscan Pro 3000 and 4000 devices Correct technique must be used to load a probe cover and ensure a secure attachment and normal operation Each shelf box contains 10 packs of 20 covers
Catalog No. 50-283-2159 Supplier Med Vet International Supplier No. 1002774BX
Thermoscan Pro 6000 probe covers are compatible for use with Thermoscan Pro 3000 and 4000 devices Correct technique must be used to load a probe cover and ensure a secure attachment and normal operation Each shelf box contains 10 packs of 20 covers