Learn More
Med Vet International McKesson Ultrasound Probe Cover, 1 X 9 Inch, 16-1007, 100/Box
Catalog No. 503044727
Encompass_Preferred
Ultrasound Probe Cover McKesson 1 X 9 Inch Polyurethane NonSterile For use with Ultrasound Probe Used as hygienic protection for ultrasound probes during ultrasound examinations Facilitates accurate ultrasound detection, while assisting in prevention of cross-contamination Straight-walled with a smooth surface and round end Non-lubricated rolled sheath
Catalog No. 50-304-4727 Supplier Med Vet International Supplier No. 1087155BX
Ultrasound Probe Cover McKesson 1 X 9 Inch Polyurethane NonSterile For use with Ultrasound Probe Used as hygienic protection for ultrasound probes during ultrasound examinations Facilitates accurate ultrasound detection, while assisting in prevention of cross-contamination Straight-walled with a smooth surface and round end Non-lubricated rolled sheath