Learn More
Wilmad Labglass COVER REAC 4N 24/40C 24/40S
Catalog No. 50806123
Encompass_Preferred
Reaction Vessel Lid, 4 Neck, Conical Flange, Center Neck Joint: 24/40, Side Neck Joint: 24/40, Flange I.D.: 4 in., and Ground for Leak-Proof Fit with a Reaction Flask
Catalog No. 50-806-123 Supplier Wilmad Labglass Supplier No. LG8031100
Reaction Vessel Lid, 4 Neck, Conical Flange, Center Neck Joint: 24/40, Side Neck Joint: 24/40, Flange I.D.: 4 in., and Ground for Leak-Proof Fit with a Reaction Flask