Learn More
Chemglass Life Sciences Cover Slip, Square, 18 x 18 mm, #1 Float Glass, 1 Oz Pkg
SDP
Catalog No. NC1998911
Encompass_Preferred
Thin square glass cover slips manufactured to #1 thickness for routine microscopy and specimen mounting. Each cover slip measures 18 x 18 mm and is supplied in 1 oz packages (approximately 242 pieces per ounce).
  • Manufactured from #1 float glass with 0.13-0.17 mm thickness.
  • Square 18 x 18 mm dimensions for small specimen coverage.
  • Sold in 1 oz packages; approximate count 242 per ounce.
  • Suitable for routine microscopy and sample mounting.
Catalog No. NC1998911 Supplier Chemglass Life Sciences Supplier No. CLS17621818
Thin square glass cover slips manufactured to #1 thickness for routine microscopy and specimen mounting. Each cover slip measures 18 x 18 mm and is supplied in 1 oz packages (approximately 242 pieces per ounce).
  • Manufactured from #1 float glass with 0.13-0.17 mm thickness.
  • Square 18 x 18 mm dimensions for small specimen coverage.
  • Sold in 1 oz packages; approximate count 242 per ounce.
  • Suitable for routine microscopy and sample mounting.