Learn More
Chemglass Life Sciences Cover Slip, Square, 25 x 25mm, #1 Float Glass, 1oz/Pkg, Approx 125/Oz
SDP
Supplier:  Chemglass Life Sciences CLS17622525
Encompass_Preferred
Cover slips are manufactured from Float Glass, #1 thickness (0.13 to 0.17mm). Sold in packages of 1oz.
Catalog No. NC1811325