Learn More
Bellco Glass, Inc. Cover Slip 9x35mm, #1 Thk
SDP
Catalog No. 501944681
Encompass_Preferred
Glass COVER SLIP Ideal for neutral tissue cell cultures and hard to attach cells. Specifically developed for fluorescence and confocal microscopy.. Provides quality, reliability, and performance for all laboratory microscopy needs. #1 thickness (0.13 to 0.16)
Catalog No. 50-194-4681 Supplier Bellco Glass, Inc. Supplier No. 191609035A
Glass COVER SLIP Ideal for neutral tissue cell cultures and hard to attach cells. Specifically developed for fluorescence and confocal microscopy.. Provides quality, reliability, and performance for all laboratory microscopy needs. #1 thickness (0.13 to 0.16)