Learn More
Sigma Aldrich Fine Chemicals Biosciences Bright-Line Hemacytometer replacement cover slip, 1 EA
Catalog No. NC0723184
Encompass
Replacement cover slip for the Bright-Line Hemacytometer: Z359629. This is the replacement cover slip for the Bright-Line Hemacytometer, Prod. No. Z359629. This product consists of a single cover slip. The microscope slide cover slips are polished soda lime glass, they are rectangular in shape and measure: 20 mm x 26 mm x 0.5 mm thick.
Catalog No. NC0723184 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3753571EA
Replacement cover slip for the Bright-Line Hemacytometer: Z359629. This is the replacement cover slip for the Bright-Line Hemacytometer, Prod. No. Z359629. This product consists of a single cover slip. The microscope slide cover slips are polished soda lime glass, they are rectangular in shape and measure: 20 mm x 26 mm x 0.5 mm thick.