Learn More
Chemglass Life Sciences Chemglass Cover Slip, Rectangular, #1.5 (24 x 50 mm), 1 oz package
SDP
Catalog No. NC2961503
Encompass_Preferred
Chemglass cover slips are rectangular microscopy cover glasses manufactured from Corning 0211 float glass with a #1.5 thickness (0.16-0.19 mm). These 24 x 50 mm cover slips are sold in 1 oz packages and are suitable for routine slide preparation and high-resolution imaging.
  • Rectangular cover slip, 24 x 50 mm.
  • Thickness #1.5 (0.16-0.19 mm).
  • Float glass material for optical clarity.
  • Sold in 1 oz packages; approximately 56 per oz.
  • Suitable for microscopy slide preparation and imaging.
Catalog No. NC2961503 Supplier Chemglass Life Sciences Supplier No. CLS17642450
Chemglass cover slips are rectangular microscopy cover glasses manufactured from Corning 0211 float glass with a #1.5 thickness (0.16-0.19 mm). These 24 x 50 mm cover slips are sold in 1 oz packages and are suitable for routine slide preparation and high-resolution imaging.
  • Rectangular cover slip, 24 x 50 mm.
  • Thickness #1.5 (0.16-0.19 mm).
  • Float glass material for optical clarity.
  • Sold in 1 oz packages; approximately 56 per oz.
  • Suitable for microscopy slide preparation and imaging.