Learn More
Chemglass Life Sciences Cover Slip, Rectangular, 9 x 35mm, #1 Float Glass, 1oz/Pkg, Approx 260/Oz
SDP
Catalog No. 50143697
Encompass_Preferred
Cover Slip, Rectangular, 9 x 35mm, #1 Float Glass, 1oz/Pkg, Approx 260/Oz. Cover slips are manufactured from Float Glass, #1 thickness (0.13 to 0.17mm). Sold in packages of 1oz.
Catalog No. 50-143-697 Supplier Chemglass Life Sciences Supplier No. CLS17620935
Cover Slip, Rectangular, 9 x 35mm, #1 Float Glass, 1oz/Pkg, Approx 260/Oz. Cover slips are manufactured from Float Glass, #1 thickness (0.13 to 0.17mm). Sold in packages of 1oz.