Learn More
Chemglass Life Sciences Cover Slip, Round, 12mm Diameter, #1 Corning 0211 Glass, 1oz/Pkg, Approx 650/Oz
SDP
Catalog No. 50143822
Encompass_Preferred
Cover slips are manufactured from Corning 0211 Float Glass, #1 thickness (0.13 to 0.16mm). Sold in packages of 1oz.
Catalog No. 50-143-822 Supplier Chemglass Life Sciences Supplier No. CLS1763012
Cover slips are manufactured from Corning 0211 Float Glass, #1 thickness (0.13 to 0.16mm). Sold in packages of 1oz.