Learn More
Chemglass Life Sciences Cover Slip, Round, 25mm Diameter, #1 Corning 0211 Glass, 1oz/Pkg, Approx 160/Oz
SDP
Catalog No. 50143784
Encompass_Preferred
Cover Slip, Round, 25mm Diameter, #1 Corning 0211 Glass, 1oz/Pkg, Approx 160/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1 thickness (0.13 to 0.16mm). Sold in packages of 1oz.5mm Diameter will fit in a 96 well plate; CLS-3500-09612mm Diameter will fit in a 24 well plate; CLS-3500-02415mm Diameter will fit in a 12 well plate; CLS-3500-01225mm Diameter will fit in a 6 well plate; CLS-3500-006
Catalog No. 50-143-784 Supplier Chemglass Life Sciences Supplier No. CLS1763025
Cover Slip, Round, 25mm Diameter, #1 Corning 0211 Glass, 1oz/Pkg, Approx 160/Oz. Cover slips are manufactured from Corning 0211 Float Glass, #1 thickness (0.13 to 0.16mm). Sold in packages of 1oz.5mm Diameter will fit in a 96 well plate; CLS-3500-09612mm Diameter will fit in a 24 well plate; CLS-3500-02415mm Diameter will fit in a 12 well plate; CLS-3500-01225mm Diameter will fit in a 6 well plate; CLS-3500-006