Learn More
Bellco Glass, Inc. Cover Slip Vac
SDP
Catalog No. 501944712
Encompass_Preferred
The cover slip vac is a small suction pick up tool for cover-slips with a 1/4" diameter tip for placing and handling. The suction cup is clear for easy viewing of cover slip that is being handled. Dimensions: 3 1/2" long
Catalog No. 50-194-4712 Supplier Bellco Glass, Inc. Supplier No. 194370000
The cover slip vac is a small suction pick up tool for cover-slips with a 1/4" diameter tip for placing and handling. The suction cup is clear for easy viewing of cover slip that is being handled. Dimensions: 3 1/2" long