Learn More
Andwin Scientific COVER SLIPPING FILM,60M-5RL/CS
SDP
Catalog No. NC9513597
Encompass_Preferred
The Sakura Finetek coverslipping film is compatible for use with Tissue-Tek SCA or Film Coverslippers. Using this film, coversliper can slide at a rate of 1080 per hour. This product can also be used as a stand alone instrument or with a Sakura Prisma Stainer. It is activated by xylene. The film is 60 millimeters in length.
Catalog No. NC9513597 Supplier Andwin Scientific Supplier No. 4770
The Sakura Finetek coverslipping film is compatible for use with Tissue-Tek SCA or Film Coverslippers. Using this film, coversliper can slide at a rate of 1080 per hour. This product can also be used as a stand alone instrument or with a Sakura Prisma Stainer. It is activated by xylene. The film is 60 millimeters in length.