Learn More
Andwin Scientific COVER SLIPS
SDP
Catalog No. NC9250887
Encompass_Preferred
The Hemacytometer cover glass is manufactured from highly polished, hard-ground glass that contains a minimum of free alkali. It has a measurement of 20 x 26 millimeters and has a thickness of 0.4 millimeters.
Catalog No. NC9250887 Supplier Andwin Scientific Supplier No. B4005
The Hemacytometer cover glass is manufactured from highly polished, hard-ground glass that contains a minimum of free alkali. It has a measurement of 20 x 26 millimeters and has a thickness of 0.4 millimeters.