Learn More
Art Robbins Instruments Cover Slips For Tersaki Trays, 100/PK
SDP
Catalog No. 501530667
Encompass
This item has a minimum qty of 2 per supplier requirements.
Glass coverslips for Terasaki trays, 50 x 75 mm, 0.19 to 0.25mm thick, 100/PK
Catalog No. 50-153-0667 Supplier Art Robbins Instruments Supplier No. 100800000
This item has a minimum qty of 2 per supplier requirements.
Glass coverslips for Terasaki trays, 50 x 75 mm, 0.19 to 0.25mm thick, 100/PK