Learn More
Med Vet International Probe Cover For Tat-2000 And Tat-5000 Temporal Thermometer, 134203, 50/Pack
Catalog No. 502829631
Encompass_Preferred
Provides hygiene and comfort during temperature checks Nested stacking inside package allows for quick, hands-free attachment Exergen probe covers have an ergonomic shape and smooth, latex-free surface Fits most Temporal Scanner™ thermometers that use disposables such as the TAT-2000 and the TAT-5000 These covers are disposable, preventing cross-contamination Covers the temperature scanner probe without diminishing accuracy
Catalog No. 50-282-9631 Supplier Med Vet International Supplier No. 762697PK
Provides hygiene and comfort during temperature checks Nested stacking inside package allows for quick, hands-free attachment Exergen probe covers have an ergonomic shape and smooth, latex-free surface Fits most Temporal Scanner™ thermometers that use disposables such as the TAT-2000 and the TAT-5000 These covers are disposable, preventing cross-contamination Covers the temperature scanner probe without diminishing accuracy